Learn More
SLC25A46 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12277-1-AP-150UL
Description
The SLC25A46 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC25A46. This antibody recognizes human, mouse, rat antigen. The SLC25A46 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
SLC25A46 | |
Polyclonal | |
Unconjugated | |
150 μL | |
91137 | |
IgG |
Western Blot, Immunofluorescence | |
333 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.