Learn More
SLC25A46 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12277-1-AP-150UL
 View more versions of this product
Product Code. 16889893
  • 4652.90 SEK / 150µL
Description
The SLC25A46 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC25A46. This antibody recognizes human, mouse, rat antigen. The SLC25A46 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
SLC25A46
Polyclonal
Unconjugated
150 μL
91137
IgG
Western Blot, Immunofluorescence
333 μg/mL
Rabbit
Primary
Human, Rat, Mouse
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SLC25A46 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL