missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC2A9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26486-1-AP-20UL
This item is not returnable.
View return policy
Description
GLUT9, or SLC2A9, is a member of the SLC2A facilitative glucose transporter family. Members of this family play a significant role in maintaining glucose homeostasis. GLUT9 may play a role in the development and survival of chondrocytes in cartilage matrices. Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
SLC2A9 | |
Polyclonal | |
Unconjugated | |
SLC2A9 | |
Glucose transporter type 9, GLUT 9, GLUT9, GLUTX, SLC2A9, UAQTL2, URATv1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117591, 56606 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NRM0 | |
SLC2A9 | |
SLC2A9 Fusion Protein Ag24135 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SLC2A9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction