missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC35A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13657-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the nucleotide-sugar transporter family. The encoded protein is a multi-pass membrane protein. It transports UDP-galactose from the cytosol into Golgi vesicles, where it serves as a glycosyl donor for the generation of glycans. Mutations in this gene cause congenital disorder of glycosylation type IIm (CDG2M). Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Specifications
SLC35A2 | |
Polyclonal | |
Unconjugated | |
SLC35A2 | |
SLC35A2, UDP Gal Tr, UDP galactose translocator, UDP galactose transporter, UGALT, UGAT, UGT, UGT1, UGT2, UGTL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100158233, 22232, 7355 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P78381, Q9R0M8 | |
SLC35A2 | |
SLC35A2 Fusion Protein Ag4524 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SLC35A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction