missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC45A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10453-1-AP-150UL
This item is not returnable.
View return policy
Description
SLC45A2 is a melanocyte differentiation antigen that is expressed in a high percentage of melanoma cell lines. A similar sequence gene in medaka, ′B, ′ encodes a transporter that mediates melanin synthesis. Mutations in this gene are a cause of oculocutaneous albinism type 4. The protein encoded by this gene encodes a melanocyte differentiation antigen that is expressed in a high percentage of melanoma cell lines. A similar sequence gene in medaka, ′B, ′ encodes a transporter that mediates melanin synthesis. Mutations in this gene are a cause of oculocutaneous albinism type 4. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
SLC45A2 | |
Polyclonal | |
Unconjugated | |
SLC45A2 | |
1A1, AIM1, MATP, Melanoma antigen AIM1, Protein AIM 1, SHEP5, SLC45A2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22293, 310152, 51151 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P58355, Q9UMX9 | |
SLC45A2 | |
SLC45A2 Fusion Protein Ag0699 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SLC45A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction