missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMN-Exon7 Mouse anti-Human, Mouse, Clone: 3A8G11, Proteintech
Mouse Monoclonal Antibody
1662.46 SEK - 4700.76 SEK
Specifications
Antigen | SMN-Exon7 |
---|---|
Clone | 3A8G11 |
Concentration | 1 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887295
|
Proteintech
60255-1-IG-20UL |
20 ÎĽL |
1662.46 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16877295
|
Proteintech
60255-1-IG-150UL |
150 ÎĽL |
4700.76 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody, 60255-1-Ig, raised against the C-terminal region (275-294 aa) encoded by the exon 7.
The SMN1 gene is part of a 500 kb inverted duplication on chromosome 5q13. This duplicated region contains at least four genes and repetitive elements which make it prone to rearrangements and deletions. The repetitiveness and complexity of the sequence have also caused difficulty in determining the organization of this genomic region. The telomeric and centromeric copies of this gene are nearly identical and encode the same protein - survival motor neuron protein. The SMN complex plays a catalyst role in the assemble of small nuclear ribonucleoproteins, the building blocks of the spliceosome. Mutations in the SMN1 gene are known to cause spinal muscular atrophy 1/2.Specifications
SMN-Exon7 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Component of gems 1, Gemin 1, SMN, SMN1, SMN2, SMNC, SMN-Exon7, SMNT, Survival motor neuron protein, T BCD541 | |
SMN1 | |
IgG1 | |
Protein G | |
Antibody |
3A8G11 | |
Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Mouse, Human | |
P97801, Q16637 | |
20595, 6606 | |
SMN-Exon7 Fusion Protein Ag16615 | |
Primary | |
-20°C | |
SMN1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SMN-Exon7 Mouse anti-Human, Mouse, Clone: 3A8G11, Proteintech