Learn More
SMTNL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14782-1-AP-20UL
This item is not returnable.
View return policy
Specifications
SMTNL2 | |
Polyclonal | |
Unconjugated | |
SMTNL2 | |
smoothelin like 2, Smoothelin like protein 2, SMTNL2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
276829, 342527, 679629 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q2TAL5, Q8CI12 | |
Smtnl2 | |
SMTNL2 Fusion Protein Ag6395 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Product Title
SMTNL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction