missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SND1 Mouse anti-Human, Mouse, Clone: 1A6A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60265-1-IG-20UL
This item is not returnable.
View return policy
Description
SND1/P100 (staphylococcal nuclease and tudor domain containing 1), also known as TudorSN, it functions in the Pim-1 regulation of Myb activity and acts as a transcriptional activatior of EBNA-2. It also interacts with EAV, NSP1,GTF2E1 and GTF2E2, and forms a ternary complex with Stat6 and POLR2A. The staphylococcal nuclease-like (SN)-domains directly interact with amino acids 1099-1758 of CBP. SND1/P100 plays an important role in the assembly of Stat6 transcriptome and stimulates IL-4-dependent transcription by mediating interaction between Stat6 and CBP.Specifications
SND1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q78PY7, Q7KZF4 | |
SND1 | |
SND1 Fusion Protein Ag1200 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG1 |
Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
1A6A4 | |
Unconjugated | |
SND1 | |
100 kDa coactivator, EBNA2 coactivator p100, p100, p100 co activator, SND1, TDRD11 | |
Mouse | |
Protein G | |
RUO | |
27044, 56463 | |
-20°C | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SND1 Mouse anti-Human, Mouse, Clone: 1A6A4, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction