missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRNP200 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23875-1-AP-20UL
This item is not returnable.
View return policy
Description
Pre-mRNA splicing is catalyzed by the spliceosome, a complex of specialized RNA and protein subunits that removes introns from a transcribed pre-mRNA segment. The spliceosome consists of small nuclear RNA proteins (snRNPs) U1, U2, U4, U5 and U6, together with approximately 80 conserved proteins. U5 snRNP contains nine specific proteins. This gene encodes one of the U5 snRNP-specific proteins. This protein belongs to the DEXH-box family of putative RNA helicases. It is a core component of U4/U6-U5 snRNPs and appears to catalyze an ATP-dependent unwinding of U4/U6 RNA duplices. Mutations in this gene cause autosomal-domit retinitis pigmentosa. Alternatively spliced transcript variants encoding different isoforms have been found, but the full-length nature of these variants has not been determined.Specifications
SNRNP200 | |
Polyclonal | |
Unconjugated | |
SNRNP200 | |
ASCC3L1, BRR2, BRR2 homolog, HELIC2, KIAA0788, SNRNP200, U5 200KD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23020, 320632 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75643, Q6P4T2 | |
Snrnp200 | |
SNRNP200 Fusion Protein Ag20748 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SNRNP200 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction