missing translation for 'onlineSavingsMsg'
Learn More

SNRPD2 Mouse anti-Human, Clone: 1F12B2, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66111-1-IG-20UL

 View more versions of this product

Product Code. 16897685

  • 1645.54 SEK / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The protein encoded by this gene belongs to the small nuclear ribonucleoprotein core protein family. It is required for pre-mRNA splicing and small nuclear ribonucleoprotein biogenesis. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

SNRPD2
Monoclonal
1.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P62316
SNRPD2
SNRPD2 Fusion Protein Ag7115
20 μL
Primary
Human
Antibody
IgG2b
Western Blot
1F12B2
Unconjugated
SNRPD2
Sm D2, SMD2, snRNP core protein D2, SNRPD2
Mouse
Protein A
RUO
6633
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SNRPD2 Mouse anti-Human, Clone: 1F12B2, Proteintech > 20 μL; Unconjugated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.