Learn More
SNRPN Mouse anti-Human, Unconjugated, Clone: 1H2F4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67280-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The SNRPN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SNRPN. This antibody recognizes human, mouse, rat antigen. The SNRPN antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
SNRPN | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Mouse, Rat, Human |
Immunohistochemistry, Western Blot | |
1H2F4 | |
Unconjugated | |
20 μL | |
6638 | |
IgG1 |