Learn More
STAT3 Rabbit, anti-Human, Unconjugated, Clone: 3H1, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand:  Proteintech 80149-1-RR-100UL
 View more versions of this product
Product Code. 17291536
  • 4982.01 SEK / 100µL
Description
Signal transducer and activator of transcription 3 (acute-phase response factor) (STAT3, synonyms: APRF, FLJ20882, MGC16063) is a member of the STAT protein family. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. STAT3 is activated through phosphorylation in response to various cytokines and growth factors including IFNs, EGF, IL5, IL6, HGF, LIF and BMP2. STAT3 mediates the expression of a variety of genes in response to cell stimuli, and thus plays a key role in many cellular processes such as cell growth and apoptosis. The small GTPase Rac1 has been shown to bind and regulate the activity of STAT3.
Specifications
Specifications
STAT3
Monoclonal
400 μg/mL
Rabbit
Primary
Human, Mouse
Western Blot, ELISA
3H1
Unconjugated
100 μL
6774
Liquid
Certificates
Promotions
Promotions
Product Content Correction
Product Title
STAT3 Rabbit, anti-Human, Unconjugated, Clone: 3H1, Proteintech > 100 μL