Learn More
Motic™ SMZ-168 Stereozoom™ Microscope
Enables precise reproduction of magnifications with 5-position click-stop magnification mechanism implemented into the continuous zoom. Motic™ SMZ168 Series Stereo Zoom Microscope combines an impressive 3-dimensional upright image with an industry leading working distance of 113mm in its standard configuration to allow easy manipulation of the sample.
11245.00 SEK - 12225.00 SEK
Specifications
Includes | Black, white and frosted stage plates and dust cover |
---|---|
Magnification Power | 0.75X to 5X |
Zoom Range | Ratio 1:6,7 |
Head Style | Rotating |
Description
- Greenough Optical System
- Achromatic optics with anti-fungus treatment
- Widefield binocular/trinocular 35° observation tube (light distribution 0:100 left eyetube/trinotube in photo position)
- Widefield high eyepoint eyepieces, WF 10x23 with diopter adjustment on both eyepiece tubes ±5dpt
- Interpupillary distance 52 to 75mm
- Zoom: 6.7:1; 5 clickstops
- 7.5X to 5X standard magnification
- Working distance: 113mm
Specifications
Black, white and frosted stage plates and dust cover | |
0.75X to 5X | |
Ratio 1:6,7 | |
Rotating |