Learn More
IBA Lifesciences Strep-Tactin™XT Spin Columns
IBA Lifesciences GmbH Strep-Tactin™XT Spin Columns allows for rapid parallel purification of Strep-tag™ and Twin-Strep-tag™ fusion proteins.
Brand: IBA Lifesciences 2-4150-025
This item is not returnable.
View return policy
Description
- Strep-Tactin™XT Spin Columns contain dried polymethacrylate, a synthetic material coupled with the streptavidin variant Strep-Tactin™XT and are suitable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
- The pre-packed spin columns are provided with receiver tubes and compatible with conventional microcentrifuges, allowing a rapid, parallel processing of multiple samples.
- Up to 400 μg of mCherry-Strep-tag™II (28 kDa) can be purified under physiological conditions in less than 10 minutes.
- Prior to application, the spin columns need to be re-hydrated and activated with Buffer W.
- In contrast to agarose resins, plant cell components such as chlorophyll are not unspecifically bound by the resin, making Strep-Tactin™XT Spin Columns a favorable alternative
- Please note that the spin columns are not reusable.
Specifications
25 Spin Columns |
Protocol Strep-TactinXT-Spin-Columns 2-4150-025
File size: 640 kB
Product Content Correction
Product Title
IBA Lifesciences Strep-Tactin™XT Spin Columns > Quantity: 25 Spin Columns