missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SULT4A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12578-1-AP-150UL
This item is not returnable.
View return policy
Description
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure(number and length of exons) is similar among family members. SULT4A1 isoforms a and b are selectively expressed in brain tissue. Isoform a is the predomit of the two isoforms of this protein. Isoform b includes an alternate segment, compared to isoform a, that causes a frameshift. The resulting protein (isoform b) has a distinct and shorter C-terminus when compared to isoform a.Specifications
SULT4A1 | |
Polyclonal | |
Unconjugated | |
SULT4A1 | |
BR STL 1, BRSTL1, hBR STL, hBR STL 1, NST, ST4A1, Sulfotransferase 4A1, SULT4A1, SULTX3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25830, 29859, 58953 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P63046, P63047, Q9BR01 | |
Sult4a1 | |
SULT4A1 Fusion Protein Ag3271 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SULT4A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction