Learn More
SUMO1 Mouse anti-Human, Unconjugated, Clone: 3B5B5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67559-1-IG-20UL
 View more versions of this product
Product Code. 16880256
  • 1645.54 SEK / 20µL
Description
The SUMO1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SUMO1. This antibody recognizes human, mouse, rat antigen. The SUMO1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
SUMO1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
3B5B5
Unconjugated
20 μL
7341
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SUMO1 Mouse anti-Human, Unconjugated, Clone: 3B5B5, Proteintech > 20 μL