missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUMO1 Mouse anti-Human, Unconjugated, Clone: 3B5B5, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | SUMO1 |
---|---|
Clone | 3B5B5 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16880256
|
Proteintech
67559-1-IG-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16870256
|
Proteintech
67559-1-IG-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SUMO1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SUMO1. This antibody recognizes human, mouse, rat antigen. The SUMO1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
SUMO1 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
7341 | |
Primary |
3B5B5 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SUMO1 Mouse anti-Human, Unconjugated, Clone: 3B5B5, Proteintech