Learn More
SUMO1 Mouse anti-Human, Unconjugated, Clone: 3B5B5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67559-1-IG-20UL
 View more versions of this product
Product Code. 16880256
  • 1645.54 SEK / 20µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
The SUMO1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SUMO1. This antibody recognizes human, mouse, rat antigen. The SUMO1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
SUMO1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
3B5B5
Unconjugated
20 μL
7341
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SUMO1 Mouse anti-Human, Unconjugated, Clone: 3B5B5, Proteintech > 20 μL