missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUPT7L Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25606-1-AP-20UL
This item is not returnable.
View return policy
Description
SUPT7L is a protein subunit of the human STAGA complex (SPT3)/TAF9/GCN5 acetyltransferase complex), which is a chromatin-modifying multiprotein complex.Specifications
SUPT7L | |
Polyclonal | |
Unconjugated | |
SUPT7L | |
Adenocarcinoma antigen ART1, ART1, KIAA0764, SPT7L, STAF65, STAF65(gamma), STAF65gamma, STAGA complex 65 subunit gamma, Suppressor of Ty 7 like, SUPT7L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9913 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O94864 | |
SUPT7L | |
SUPT7L Fusion Protein Ag22264 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SUPT7L Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction