Learn More
Synaptogyrin-4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21751-1-AP-150UL
 View more versions of this product
Product Code. 16881345
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Synaptogyrins comprise a family of tyrosine-phosphorylated membrane proteins with two neuronal (SYNGR1 and SYNGR3) and one ubiquitous (SYNGR2) members. SYNGR1 and -3 are synaptic vesicle proteins, residing in some cases on the same synaptic vesicle, and are thought to be involved in the regulation of neurotransmitter release. SYNGR2, by contrast, is absent from synaptic vesicles. SYNGR3 is predomitly expressed in brain and placenta. The role and localization of a fourth synaptogyrin, SYNGR4, is unclear.
Specifications
Specifications
Synaptogyrin-4
Polyclonal
Unconjugated
SYNGR4
synaptogyrin 4, Synaptogyrin-4, SYNGR4
Rabbit
Antigen Affinity Chromatography
RUO
23546, 292916, 58867
-20°C
Liquid
Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95473, Q9Z1L2
Syngr4
Synaptogyrin-4 Fusion Protein Ag15865
150 ÎĽL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Synaptogyrin-4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 ÎĽL; Unconjugated