Learn More
Thermo Scientific™ Tango™ CXCR2-bla U2OS Cells
The Tango™ CXCR2-bla U2OS cells contain the Chemokine (C-X-C motif) Receptor 2 (CXCR2) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
Brand: Thermo Scientific™ K1521
Description
The Tango™ CXCR2-bla U2OS cells contain the Chemokine (C-X-C motif) Receptor 2 (CXCR2) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
The Tango™ CXCR2-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of established ligands. In addition, Tango™ CXCR2-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product.
Specifications
NM_001557.2 | |
Dividing Cells | |
Tail from GRPR GPCR | |
CXCR2 | |
GeneBLAzer™ | |
8 x 106 Cells | |
BLA (Beta-Lactamase) | |
U2OS | |
GPCR |
Cell-based beta-arrestin recruitment | |
(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • Tango™ CXCR2-bla U2OS cells • Tango™ CXCR2-bla U2OS Assay Protocol • Certificate of Analysis The Tango™ CXCR2-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
|
Fluorescence | |
Pool of Stable Cells | |
GPCR (G-Protein Coupled Receptors) | |
End Point | |
CXCR2 | |
Tango™ |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1860685 | Certificate of Analysis | 19/07/2017 | K1521 |