Learn More
Thermo Scientific™ Tango™ GPR39-bla U2OS Cells
Tango™ GPR39-bla U2OS cells contain the orphan human G-protein Coupled Receptor 39 (GPR39) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
Brand: Thermo Scientific™
Description
Tango™ GPR39-bla U2OS cells contain the orphan human G-protein Coupled Receptor 39 (GPR39) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase reporter gene under the control of a UAS response element. The Tango™ GPR39-bla U2OS cells have been functionally validated for a receptor specific response to FBS. In addition, Tango™ GPR39-bla U2OS cells have been tested for assay performance under variable conditions.
Specifications
NM_001508.1 | |
Dividing Cells | |
Native Tail | |
FRET | |
G-Protein Coupled Receptor (GPCR) | |
Pool of Stable Cells | |
8 x 106 Cells | |
GPR39 | |
U2OS | |
GPCR |
Cell-based beta-arrestin recruitment | |
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • Tango™ GPR39-UAS-bla CHO-K1 cells • Tango™ GPR39-UAS-bla CHO-K1 Assay Protocol • Certificate of Analysis The Tango™ GPR39-UAS-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
|
Fluorescence | |
GPR39 | |
GPR39-UAS-bla CHO-K1 cells, GPR39-UAS-bla CHO-K1 Assay Protocol, Certificate of Analysis | |
Tango™ | |
End Point | |
Dividing Cells | |
Tango™ |