Learn More
Tantalum Solution for ICP, 1g/L, HNO3/HF Matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
762.00 SEK - 1710.00 SEK
Specifications
Solution Contains | HNO3 |
---|---|
Concentration | 1 g/L |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12983514
![]() |
Fisher Chemical
S 057 100 |
100 mL |
762.00 SEK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12993514
![]() |
Fisher Chemical
S 057 250 |
250 mL |
1140.00 SEK
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12903524
![]() |
Fisher Chemical
S 057 500 |
500 mL |
1710.00 SEK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
HNO3 | |
1 g/L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Single-element Standard Solution for ICP. Tantalum (Ta) 1000mg/l in HNO3 5%,HF 1%
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Tantalum Solution for ICP, 1g/L, HNO3/HF Matrix, Fisher Chemical™