missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBRG4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16245-1-AP-150UL
This item is not returnable.
View return policy
Description
TBRG4 (transforming growth factor beta regulator 4), also known as CPR2 (cell cycle progression restoration protein 2) or FAST 4 (FAST kinase domain-containing protein 4), is a 631 amino acid protein that contains one RAP domain and belongs to the FAST kinase family. TBRG4 is ubiquitously expressed and may have a role in cell cycle progression. Existing as two alternatively spliced isoforms, the gene encoding TBRG4 maps to human chromosome 7p13.Specifications
TBRG4 | |
Polyclonal | |
Unconjugated | |
TBRG4 | |
CPR2, FASTKD4, KIAA0948, Protein TBRG4, TBRG4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21379, 360977, 9238 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5M9G9, Q91YM4, Q969Z0 | |
Tbrg4 | |
TBRG4 Fusion Protein Ag9176 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TBRG4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction