missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TC-S 7009
GREENER_CHOICE

High affinity and selective HIF-2α inhibitor

1320.00 SEK - 4430.00 SEK

Specifications

Chemical Name or Material N-(3-Chloro-5-fluorophenyl)-4-nitro-2,1,3-benzoxadiazol-5-amine
CAS 1422955-31-4
Assay Percent Range >99%
Molecular Formula C12H6ClFN4O3
Formula Weight Observed MW: 308.65
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18747418
GREENER_CHOICE
View Documents
Tocris Bioscience™
5243/25
25 mg
4430.00 SEK
25mg
Please to purchase this item. Need a web account? Register with us today!
18771488
GREENER_CHOICE
View Documents
Tocris Bioscience™
5243/5
5 mg
1320.00 SEK
5mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

High affinity and selective HIF-2α inhibitor (Kd = 81 nM). Binds to the HIF-2α PAS-B domain; disrupts HIF-2α-ARNT heterodimerization. Exhibits >60-fold selectivity for HIF-2α over HIF-1α. Decreases HIF-2α DNA-binding and attenuates expression of HIF-2α target genes in vitro.
Specifications

Specifications

N-(3-Chloro-5-fluorophenyl)-4-nitro-2,1,3-benzoxadiazol-5-amine
>99%
Observed MW: 308.65
1422955-31-4
C12H6ClFN4O3
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at Room Temperature

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tocris Bioscience™ TC-S 7009

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.