Learn More
TCF7L1/TCF7L2 Mouse anti-Human, Unconjugated, Clone: 2B2D12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67652-1-IG-20UL
Description
The TCF7L1/TCF7L2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TCF7L1/TCF7L2. This antibody recognizes human, mouse antigen. The TCF7L1/TCF7L2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
TCF7L1/TCF7L2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse |
Western Blot, ELISA | |
2B2D12 | |
Unconjugated | |
20 μL | |
83439 | |
IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.