Learn More
TCF7L1/TCF7L2 Mouse anti-Human, Unconjugated, Clone: 2B2D12, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TCF7L1/TCF7L2 |
---|---|
Clone | 2B2D12 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16800406
|
Proteintech
67652-1-IG-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16890396
|
Proteintech
67652-1-IG-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TCF7L1/TCF7L2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TCF7L1/TCF7L2. This antibody recognizes human, mouse antigen. The TCF7L1/TCF7L2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
TCF7L1/TCF7L2 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
83439 | |
Primary |
2B2D12 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Mouse | |
IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.