Learn More
TCF7L1/TCF7L2 Mouse anti-Human, Unconjugated, Clone: 2B2D12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67652-1-IG-150UL
 View more versions of this product
Product Code. 16890396
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The TCF7L1/TCF7L2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TCF7L1/TCF7L2. This antibody recognizes human, mouse antigen. The TCF7L1/TCF7L2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TCF7L1/TCF7L2
Monoclonal
1000 ÎĽg/mL
Mouse
Primary
Human, Mouse
Western Blot, ELISA
2B2D12
Unconjugated
150 ÎĽL
83439
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TCF7L1/TCF7L2 Mouse anti-Human, Unconjugated, Clone: 2B2D12, Proteintech > 150 ÎĽL