missing translation for 'onlineSavingsMsg'
Learn More

TCN2 Mouse anti-Human, Clone: 2G6D2, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66377-1-IG-150UL

 View more versions of this product

Product Code. 16888195

  • 4652.90 SEK / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Transcobalamin I (TCI) and Transcobalamin II (TCII) are secreted proteins belonging to the eukaryotic cobalamin transport proteins family and also to the vitamin B12-binding protein family. The genes encoding these proteins map to chromosome 11q11-q12 and 22q12.2, respectively. Transcobalamin I is a constituent of secondary granules in neutrophils, while Transcobalamin II binds cobalamin and mediates its transport into cells. These plasma proteins are expressed in various tissues and secretions.
Specifications

Specifications

TCN2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P20062
TCN2
TCN2 Fusion Protein Ag2825
150 μL
Primary
Human
Antibody
IgG2b
Western Blot
2G6D2
Unconjugated
TCN2
TC 2, TC II, TC2, TCII, TCN2, Transcobalamin 2, Transcobalamin II
Mouse
Protein A
RUO
6948
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TCN2 Mouse anti-Human, Clone: 2G6D2, Proteintech > 150 μL; Unconjugated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.