missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCN2 Mouse anti-Human, Clone: 2G6D2, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TCN2 |
---|---|
Clone | 2G6D2 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16898195
|
Proteintech
66377-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16888195
|
Proteintech
66377-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Transcobalamin I (TCI) and Transcobalamin II (TCII) are secreted proteins belonging to the eukaryotic cobalamin transport proteins family and also to the vitamin B12-binding protein family. The genes encoding these proteins map to chromosome 11q11-q12 and 22q12.2, respectively. Transcobalamin I is a constituent of secondary granules in neutrophils, while Transcobalamin II binds cobalamin and mediates its transport into cells. These plasma proteins are expressed in various tissues and secretions.Specifications
TCN2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
TC 2, TC II, TC2, TCII, TCN2, Transcobalamin 2, Transcobalamin II | |
TCN2 | |
IgG2b | |
Protein A | |
Antibody |
2G6D2 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P20062 | |
6948 | |
TCN2 Fusion Protein Ag2825 | |
Primary | |
-20°C | |
TCN2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TCN2 Mouse anti-Human, Clone: 2G6D2, Proteintech