missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP10L Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13709-1-AP-20UL
This item is not returnable.
View return policy
Description
TCP10L is a human leucine zipper protein with transcription inhibition activity.Specifications
TCP10L | |
Polyclonal | |
Unconjugated | |
TCP10L | |
C21orf77, PRED77, t complex 10 (mouse) like, T complex protein 10A 2, TCP10 like, TCP10A 2, TCP10L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140290 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8TDR4 | |
TCP10L | |
TCP10L Fusion Protein Ag4646 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TCP10L Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction