missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP10L Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TCP10L |
---|---|
Concentration | 0.34 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16842064
|
Proteintech
13709-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16832064
|
Proteintech
13709-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCP10L is a human leucine zipper protein with transcription inhibition activity.Specifications
TCP10L | |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8TDR4 | |
140290 | |
TCP10L Fusion Protein Ag4646 | |
Primary | |
-20°C | |
TCP10L |
0.34 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C21orf77, PRED77, t complex 10 (mouse) like, T complex protein 10A 2, TCP10 like, TCP10A 2, TCP10L | |
TCP10L | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TCP10L Rabbit anti-Human, Mouse, Polyclonal, Proteintech