missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDP-43 Mouse anti-Human, Mouse, Rat, Clone: 1G4F5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66734-1-IG-150UL
This item is not returnable.
View return policy
Description
66734-1-Ig is a mouse monoclonal antibody recognizing the intact protein of TDP-43.
Various biochemical, physiological and behavioural processes display circadian rhythms controlled by an internal biological clock. The central "gears" driving this clock appear to be composed of an autoregulatory transcription/posttranslation-based feedback loop. Cryptochrome 1 (CRY1) and 2 (CRY2) are DNA-binding flavoproteins that bear some homology to blue-light receptors and photolyases. In Drosophila, CRY is a photoreceptor for the circadian clock where it binds to the clock component TIM in a light-dependent fashion and blocks its function. Mammalian CRY1 and CRY2 function via light-independent interactions with circadian genes CLOCK and BMAL1, as well as with PER1, PER2, and TIM. They seem to act as light-independent components of the circadian clock and likely regulate Per1 transcriptional cycling via interactions with both the activator and its feedback inhibitors. Mutant mice not expressing the Cry1 or Cry2 protein display accelerated and delayed periodicity of locomotor activity, respectively. It appears that the combination of both proteins working together is essential to synchronize the organism to circadian phases. A critical balance between Cry1 and Cry2 is required for proper clock function; in complete darkness, double-mutant mice present with instantaneous arrhythmicity, indicating the absence of an internal circadian clock.Specifications
TDP-43 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13148, Q921F2 | |
Tardbp | |
TDP-43 Fusion Protein Ag1231 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1G4F5 | |
Unconjugated | |
TARDBP | |
ALS10, TAR DNA binding protein, TAR DNA binding protein 43, TARDBP, TDP 43, TDP43 | |
Mouse | |
Protein G | |
RUO | |
230908, 23435, 298648 | |
-20°C | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TDP-43 Mouse anti-Human, Mouse, Rat, Clone: 1G4F5, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction