missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEX38 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24429-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody detects 25-28 kDa with mouse testis and MDA-MB-453s cell lysate.
TEX38 is a protein coding gene.Specifications
TEX38 | |
Polyclonal | |
Unconjugated | |
TEX38 | |
ATPAF1-AS1, C1orf223, TEX38 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
374973, 75173 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A2A8T7, Q6PEX7 | |
TEX38 | |
TEX38 Fusion Protein Ag19833 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TEX38 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction