Learn More
TFPT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10097-2-AP-20UL
 View more versions of this product
Product Code. 16886253
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
TFPT component of the chromatin-remodeling INO80 complex which, in addition to chromatin remodeling implicated in crucial functions such as DNA repair, checkpoint regulation, DNA replication, telomemere maintece and chromosome segregation. This protein appears to promote apoptosis in a TP53/p53-independent manner.
Specifications
Specifications
TFPT
Polyclonal
Unconjugated
TFPT
amida, FB1, INO80 complex subunit F, INO80F, Protein FB1, TCF3 fusion partner, TFPT
Rabbit
Antigen Affinity Chromatography
RUO
29844, 69714, 85423
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.42 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P0C1Z6, Q3U1J1, Q9JMG6
Tfpt
TFPT Fusion Protein Ag0131
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TFPT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated