missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGDS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17310-1-AP-20UL
This item is not returnable.
View return policy
Specifications
TGDS | |
Polyclonal | |
Unconjugated | |
TGDS | |
dTDP D glucose 4,6 dehydratase, SDR2E1, TDP glucose 4,6 dehydratase, TDPGD, TGDS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23483, 306164, 76355 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95455, Q8VDR7 | |
Tgds | |
TGDS Fusion Protein Ag11276 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TGDS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu