missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGDS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TGDS |
---|---|
Concentration | 0.21 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887174
|
Proteintech
17310-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16877174
|
Proteintech
17310-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
TGDS | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
O95455, Q8VDR7 | |
23483, 306164, 76355 | |
TGDS Fusion Protein Ag11276 | |
Primary | |
-20°C | |
TGDS |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
dTDP D glucose 4,6 dehydratase, SDR2E1, TDP glucose 4,6 dehydratase, TDPGD, TGDS | |
Tgds | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TGDS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech