missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGN46 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13573-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
IASPP is one of the most evolutionarily conserved inhibitors of p53 (TP53; MIM 191170), whereas ASPP1. and ASPP2. are activators of p53.Specifications
TGN46 | |
Polyclonal | |
Unconjugated | |
TGOLN2 | |
Tgn38, TGN38 homolog, TGN46, TGN48, TGN51, TGOLN2, trans Golgi network protein 2, TTGN2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10618 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43493 | |
TGOLN2 | |
TGN46 Fusion Protein Ag4470 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |