Learn More
MedChemExpress Thalidomide-O-amido-PEG2-C2-NH2 TFA
MedChemExpress Thalidomide-O-amido-PEG2-C2-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and 2-unit PEG linker used in PROTAC technology.

POA - 28133.57 SEK
Specifications
Molecular Weight (g/mol) | 576.48 g/mol |
---|---|
CAS | 1957235-75-4 |
Content And Storage | 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture) |
Shipping Condition | Room temperature |
Synonym | Cereblon Ligand-Linker Conjugates 10 TFA ∣ E3 Ligase Ligand-Linker Conjugates 24 TFA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16491606
![]() |
MedChemExpress
HY-112617A |
500 mg |
28133.57 SEK
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16471179
![]() |
MedChemExpress
HY-112617A |
1 g |
POA
1g |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16465496
![]() |
MedChemExpress
HY-112617A |
100 mg |
7873.67 SEK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16440146
![]() |
MedChemExpress
HY-112617A |
2 g |
POA
2g |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
576.48 g/mol | |
4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture) | |
Cereblon Ligand-Linker Conjugates 10 TFA ∣ E3 Ligase Ligand-Linker Conjugates 24 TFA |
1957235-75-4 | |
Room temperature | |
H2O : 150 mg/mL (260.20 mM; Need ultrasonic) |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.