Learn More
Thermo Scientific™ Nalgene™ Replacement Closures for Economy Wash Bottles
GREENER_CHOICE
Through-the-top tubing with closure and stem molded in one piece
1445.00 SEK - 1455.00 SEK
Specifications
Material Polypropylene
For Use With (Equipment) Wash bottle
View More Specs
Products 2
Product Code Brand Neck Finish Price Quantity & Availability  
Product Code Brand Neck Finish Price Quantity & Availability  
10385651
GREENER_CHOICE
View Documents
Thermo Scientific™
712401-0002
24-400
1455.00 SEK
Pack of 25
Only null left
Estimated Shipment: 21-11-2025
Add to basket
10050561
GREENER_CHOICE
View Documents
Thermo Scientific™
712401-0001
28-400
1445.00 SEK
Pack of 25
Only null left
In Stock
Add to basket
Description
Description
  • Closure and stem molded in one piece for leakproof service
  • Tip can be cut back to provide increased flow
  • Tight fit of draw tube and closure prevents separation when squeezing bottle
  • Leakproof†
Specifications
Specifications
Polypropylene
Wash bottle
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Replacement Closures for Economy Wash Bottles