missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THUMPD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14921-1-AP-150UL
This item is not returnable.
View return policy
Description
Functions as a tRNA-binding adapter to mediate NAT10-dependent tRNA acetylation.Specifications
THUMPD1 | |
Polyclonal | |
Unconjugated | |
THUMPD1 | |
THUMP domain containing 1, THUMPD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
233802, 309041, 55623 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99J36, Q9NXG2 | |
Thumpd1 | |
THUMPD1 Fusion Protein Ag6739 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
THUMPD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction