missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THUMPD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | THUMPD1 |
---|---|
Concentration | 0.15 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883834
|
Proteintech
14921-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873834
|
Proteintech
14921-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Functions as a tRNA-binding adapter to mediate NAT10-dependent tRNA acetylation.Specifications
THUMPD1 | |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q99J36, Q9NXG2 | |
233802, 309041, 55623 | |
THUMPD1 Fusion Protein Ag6739 | |
Primary | |
-20°C | |
THUMPD1 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
THUMP domain containing 1, THUMPD1 | |
Thumpd1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
THUMPD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech