missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymidine Phosphorylase / PD-ECGF Mouse, Clone: P-GF.44C, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 1890-MSM1-P0
This item is not returnable.
View return policy
Specifications
Thymidine Phosphorylase / PD-ECGF | |
Cytoplasmic and Nuclear | |
P-GF.44C | |
Unconjugated | |
NP_001107227 | |
Mouse | |
55kDa | |
Protein A/G Purified | |
RUO | |
1890 | |
IgG1 |
Immunohistochemistry | |
Monoclonal | |
200ÎĽg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
TYMP | |
Recombinant full-length human Thymidine Phosphorylase (TP / PD-ECGF) protein | |
HUVEC cells. Breast, Bladder, Lung or Kaposi tumors | |
20 ÎĽg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thymidine Phosphorylase / PD-ECGF Mouse, Clone: P-GF.44C, enQuireBio™ > 20 μg
Spot an opportunity for improvement?Share a Content Correction