missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymidine Phosphorylase / PD-ECGF Mouse, Clone: P-GF.44C, enQuireBio™
Mouse Monoclonal Antibody
2035.00 SEK - 5965.00 SEK
Specifications
Antigen | Thymidine Phosphorylase / PD-ECGF |
---|---|
Clone | P-GF.44C |
Concentration | 200ÎĽg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15989907
|
EnquireBio
1890-MSM1-P0 |
20 ÎĽg | N/A | ||||||
15999907
|
EnquireBio
1890-MSM1-P1 |
100 ÎĽg |
5965.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Thymidine Phosphorylase / PD-ECGF | |
200ÎĽg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
1890 | |
Recombinant full-length human Thymidine Phosphorylase (TP / PD-ECGF) protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
55kDa |
P-GF.44C | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_001107227 | |
TYMP | |
IgG1 | |
Protein A/G Purified | |
Cytoplasmic and Nuclear | |
HUVEC cells. Breast, Bladder, Lung or Kaposi tumors |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thymidine Phosphorylase / PD-ECGF Mouse, Clone: P-GF.44C, enQuireBio™