missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tim23 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11123-1-AP-150UL
This item is not returnable.
View return policy
Description
Essential component of the TIM23 complex, a complex that mediates the translocation of transit peptide-containing proteins across the mitochondrial inner membrane.Specifications
Tim23 | |
Polyclonal | |
Unconjugated | |
TIMM23 | |
PRO1197, TIM23, TIMM23 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100287932, 53600, 54312 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14925, O35093, Q9WTQ8 | |
Timm23 | |
Tim23 Fusion Protein Ag1620 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tim23 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction