missing translation for 'onlineSavingsMsg'
Learn More

TIMM9 Recombinant Protein, Abnova

3860.70 SEK - 5854.80 SEK

Specifications

Cross Reactivity Human
Format Solution
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 35.53
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16108362
View Documents Promotion Details
Abnova
H00026520-P01.10UG
10 ug
3860.70 SEK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-03-2022
Please to purchase this item. Need a web account? Register with us today!
16118362
View Documents Promotion Details
Abnova
H00026520-P01.25UG
25 ug
5854.80 SEK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-03-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

Human
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
35.53
8
Glutathione Sepharose 4 Fast Flow
Yes
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Solution
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
TIMM9 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
SDS
Product Certifications