Learn More
Titanium solution for ICP 10g/L HNO3/HF matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
1215.00 SEK - 4110.00 SEK
Specifications
Analyte or Component Names | Ti |
---|---|
Concentration | 10±0.02 g/L |
Type | Mass Spectrometry Standard |
Element | Ti |
Solvent or Matrix | HCl/HF |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
12936424
![]() |
Fisher Chemical
S 261 100 |
100 mL | Bottle |
1215.00 SEK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12946424
|
Fisher Chemical
S 261 250 |
250 mL | Bottle | N/A | ||||||
12956424
|
Fisher Chemical
S 261 500 |
500 mL | Bottle | N/A | ||||||
Specifications
Ti | |
Mass Spectrometry Standard | |
HCl/HF |
10±0.02 g/L | |
Ti |
Safety and Handling
H225; H304; H315; H336; H410
P210; P233; P240; P241; P261; P264; P271; P273; P280; P301+P310; P302+P352; P303+P361+P353; P304+P340; P312; P321; P331; P332+P313; P370+P378; P391; P403+P233; P403 +P235; P405; P501
Danger
missing translation for 'shelfLife' : 24 months (12 months after opening)
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Single-element Standard Solution for ICP. Titanium (Ti) 10000mg/l in HCl 5%,HF 0.5%
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Titanium solution for ICP 10g/L HNO3/HF matrix, Fisher chemical™