missing translation for 'onlineSavingsMsg'
Learn More

Titanium solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™
BIOPHARMA

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

684.00 SEK - 735.00 SEK

Chemical Identifiers

CAS 7440-28-0
Molecular Formula Tl
Molecular Weight (g/mol) 204.38
MDL Number MFCD00134063
InChI Key BKVIYDNLLOSFOA-UHFFFAOYSA-N
PubChem CID 5359464
ChEBI CHEBI:30440
IUPAC Name thallium
SMILES [Tl]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
12977433
View Documents
Fisher Chemical
S 662 050
50 mL Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12987433
BIOPHARMA
View Documents
Fisher Chemical
S 662 100
100 mL Bottle
684.00 SEK
100mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 12 months

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.