Learn More
TMED4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14141-1-AP-150UL
 View more versions of this product
Product Code. 16882644
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Involved in endoplasmic reticulum stress response. May play a role in the regulation of heat-shock response and apoptosis.
Specifications
Specifications
TMED4
Polyclonal
Unconjugated
TMED4
ERS25, HNLF, TMED4
Rabbit
Antigen Affinity Chromatography
RUO
103694, 222068, 305502
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7Z7H5, Q8R1V4
Tmed4
TMED4 Fusion Protein Ag5234
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TMED4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated