missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMED4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TMED4 |
---|---|
Concentration | 0.15 mg/mL |
Applications | Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16892644
|
Proteintech
14141-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16882644
|
Proteintech
14141-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Involved in endoplasmic reticulum stress response. May play a role in the regulation of heat-shock response and apoptosis.Specifications
TMED4 | |
Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q7Z7H5, Q8R1V4 | |
103694, 222068, 305502 | |
TMED4 Fusion Protein Ag5234 | |
Primary | |
-20°C | |
TMED4 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ERS25, HNLF, TMED4 | |
Tmed4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TMED4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech